NEBNext® reagents for NGS sample preparation
Library preparation is a critical part of the next generation sequencing workflow; successful sequencing requires the generation of high quality libraries of sufficient yield and quality from ever-decreasing input quantities as well as from samples of lower quality. At the same time, there is a need for faster, automatable protocols that perform reliably without compromising the quality of the libraries produced.
To meet these growing challenges, the NEBNext suite of products continues to evolve to support next generation sequencing with sample preparation tools that streamline workflows, minimize inputs, and improve library yields and quality.
NEBNext Ultra™ II FS DNA Library Prep for Illumina
NEBNext Ultra II FS utilizes novel enzymatic DNA fragmentation integrated into library prep, and offers high quality, high yield library preparation with minimal input.
NEBNext Ultra II Directional RNA Library Prep for Illumina
Even more from less - now for RNA
NEBNext Multiplex Oligos
Highly pure adaptors and primers for multiplexing, including unique dual indices.