Esteem™ Comfort Nitrile Exam Gloves, Cardinal
Supplier: Avantor
76311-742BX 92.17 CAD
76311-742 76311-744 76311-746 76311-752 76311-754
Esteem™ Comfort Nitrile Exam Gloves, Cardinal
Gloves
Esteem™ Comfort are the newest generation of Esteem™ stretchy nitrile exam gloves.
  • 30% higher minimum elongation than the minimum ASTM standard for a nitrile exam glove
  • Reduce cuff tears during donning
  • Reduces the risk of developing a Type IV hypersensitivity to chemical accelerants
  • Tested against chemotherapy drugs per ASTM D6978
  • Tested against 18 chemicals
  • Textured fingertips for enhanced grip
These gloves offer the perfect combination of improved elongation with outstanding protection from chemotherapy drugs, and an FDA-approved low dermatitisMore Product Information