Cloning & Synthetic Biology
Built on over 40 years of experience, New England Biolabs provides you with the quality reagents and technical support that you need to take your cloning experiments to the next level. With reagents for each step in the cloning workflow, NEB products deliver quality and performance so that you can Clone with Confidence.
Cloning Competent E.coli from NEB
Ensure successful transformations with NEB competent E. coli for cloning. Choose from several high efficiency competent cell strains in a variety of formats that deliver performance, convenience and value. High transformation efficiences.All strains are f
NEBuilder® HiFi DNA Assembly
NEBuilder HiFi DNA Assembly enables virtually error-free joining of DNA fragments, even those with 5´- and 3´-end mismatches.
Restriction Enzymes from NEB
With >285 restriction enzymes that are 100% active in a single buffer, CutSmart, it is much simpler to set up your double digest reactions.
Q5 Site-Directed Mutagenesis Kit (available with or without competent cells)
The Q5 Site-Directed Mutagenesis Kit enables rapid, site-specific mutagenesis of double-stranded plasmid DNA in less than 2 hours.