Avantor Seradigm Sera
Cell culture experts everywhere are discovering how well their cells thrive on nutritionally superior sera from Avantor Seradigm. Seradigm’s collection and manufacturing techniques preserve more of the naturally-occurring growth factors in sera that cells require to thrive. Our proven track record for performance and consistency enables sera users to switch to a brand that yields more of the results they seek – no matter the application, scale, or budget.
Request Information / SamplesCells Choose Seradigm
The unique collection and manufacturing techniques used for our sera preserve more of the naturally-occurring growth factors that cells require to thrive. Our proven track record for performance and consistency enables sera users to switch to a brand that yields more of the results they seek – no matter the application, scale, or budget.
- Nutritionally Superior Sera
- Proven Performance and Consistency
- Supply Stability with industry-leading traceability and quality
- Unmatched operational support and service
Avantor offers a trusted supply-chain of sera with…
Quality & choice
- Proven sera performance & consistency
- Proprietary manufacturing methods result in lot-to-lot and bottle-to-bottle consistency
- Quality you can see in the crystal clear, vibrant color of each bottle of sera
Expertise
- Nutritionally superior sera
- Unique collection & manufacturing techniques preserve more naturally-occurring growth factors found in serum
- Industry-leading traceability & quality
- Independent labs used for all lot release criteria
- ISIA traceability certification confirms truth in labeling
Convenience
- Strong supply partnerships and a vertically-integrated raw material supply chain provide long-term assurance of supply
Customization
- Unmatched operational sera support & service
- Flexibility in meeting each customer’s unique needs
- Multiple lot sampling, large volume reserves, and storage solutions
- Proven track record of operational excellence, support, and on-time delivery
![]() |
We proudly feature a fully certified supply chain for traceability by the International Serum Industry Association (ISIA). For additional information, visit www.serumindustry.org |
We have established a trusted partnership with Oritain for independent origin verification testing on select FBS products. For additional information visit www.oritain.com.
Product Selection
Australia Origin FBS
Australia Origin FBS combines outstanding performance and quality with ease of regulatory access globally, suitable for a variety of applications.
Premium Grade FBS
Premium Grade FBS is widely used by cell culturists requiring high quality, high performance FBS for a variety of applications.
Ultimate Grade FBS
Ultimate Grade FBS is the ultimate in product quality and viral safety, undergoing additional BVD virus testing using qPCR technology.
Ultra Low IgG FBS
Ultra Low IgG FBS meets a specification of ≤ 5 μg/mL for applications where naturally occurring levels of IgG are too high.
Select Grade FBS
Select Grade FBS is an affordable, quality product providing consistent performance for cost-sensitive applications.