High Performance Liquid Chromatography
High performance liquid chromatography (HPLC) is an analytical technique to separate, identify, and quantify components in a mixture. It is the single biggest chromatography technique essential to most laboratories worldwide.
Featured Suppliers:
Thermo Scientific Chromatography
Comprehensive range of columns and consumables to achieve repeatable, predictable results – separation after separation.
AMT HPLC Columns
AMT’s HPLC columns are manufactured using Fused-Core® particle technology specially developed to deliver hyper-fast liquid chromatographic separations for small and large molecules.
HPLC Solvent Solutions by SCAT
SCAT products block vapors, keep your solvent clean, and filter the air exchange from waste canisters. They protect your health, the environment, and the quality of your analysis.
Featured Products
Acetonitrile HPLC, VWRC- BDH
From the exclusive family of high-quality, competitively priced BDH Chemicals– Super gradient grade for HPLC. Filtered through a 0.2 µm filter, packaged under nitrogen.
Avantor HPLC Columns
Selecting the correct column is key to your success.There are thousands of columns to chose from. We are here to help you find the right column for your needs.
Sampling
Products:
Sample Storage & Handling
Products:
Sample Preparation
Analysis
Results, Reviews & Data Storage
What is HPLC?
High performance liquid chromatography, known as HPLC, works by using high-pressure pumps to effectively force a sample mixture containing the solvent through a column that contains a solid absorbent material.
This absorbent material interacts differently with each component in the solvent. As the solvent components flow out of the column, they flow at a unique rate.
The difference in flow rate lets each component be separated from the mixture. A detector then determines the identity and concentration of the compounds.