HALYARD* Cleanroom & Life Science PPE
Cleanroom and Life Science professionals need suppliers they can depend on to deliver high-quality products that meet their specific needs. Owens & Minor, a leader in global healthcare solutions, offers a comprehensive portfolio of Personal Protective Equipment (PPE), including HALYARD* branded PUREZERO* Cleanroom and Life Science Nitrile Gloves and Masks. The HALYARD* product brand produces and supplies PPE that lead with the company’s quality and innovation, which stems from 140-plus years of industry expertise and supply reliability.
PUREZERO* Cleanroom Gloves
Accelerator-free1 portfolio offered in three non-sterile and two sterile cleanroom gloves to address the needs of pharmaceutical, medical device, microelectronics and semiconductor manufacturing.
*1 Not fromulated with these commonly used vulcanizing chemicals: Sulfur, Thiurams, Thiaxoles, Guanidines and Carbonates
Other PPE and Sterilization Wrap
The HALYARD* PPE portfolio includes a full line for both staff and patients. HALYARD* Sterilization Wrap is offered in a variety weights and sizes.