Azure Biosystems Western Blotting
Azure Biosystems offers top-of-the-line reagents and imaging systems for an efficient Western blotting workflow. With unique and flexible imaging systems supported by a range of reagents and supplies, Azure can get you high-quality data and confidence in your results. Chemiluminescence or fluorescence, gel imaging or three-color Western blotting, or our new biomolecular imager – let Azure help you each step of the way.
Azure Gel Imaging Systems
Trial the Azure Cielo for 7 Days
The Azure Cielo™ 6 Real-Time PCR system brings you the accuracy and sensitivity you need for your research, with intuitive touch screen software.
Azure Sapphire™ Biomolecular Imager
The next generation lab imaging platform supports long & short wavelengths of near infrared fluorescence (NIR), red/green/blue (RGB) imaging, chemiluminescent imaging, phosphor imaging as well as optical densitometry (OD) of proteins in stained gels.
Azure Advanced Imaging Systems
High performance imaging capabilities into a bench-friendly footprint. Choose based on applications and needs; proof that quality can be affordable.
Azure Gel Imaging Systems
Azure 200 allows you to image DNA and protein gels without a darkroom. Fully upgradable to add fluorescent or chemiluminescent Western detection