Chester Manufacturing Facility
Quality Control lab
Returnable Containers
Bulk tankers
Core Capabilities
- Production of HPLC and other high-purity grades of acetonitrile and methanol for laboratory and biopharmaceutical applications
- Packaging of HPLC acetonitrile and methanol into bulk tankers and returnable containers
- Proprietary purification process equipment developed on-site
- Closed-system manufacturing processes
- Polishing of lab and process chemicals to remove trace impurities
- Expertise in handling high-purity material while maintaining product integrity
- Support for customer use of pressurized containers and pressure dispense systems
Industry Segments and Applications
- Pharmaceutical
- Biotechnology
- Agroscience
- High-Purity Analytical Applications
Major Products
- Ultra high-purity bulk acetonitrile
- Ultra high-purity acetonitrile and methanol in recyclable containers
- Commercial grade acetonitrile and methanol in bulk, recyclable container and drum format
Quality Systems
- ISO 9001
- Propriety quality system designed to meet evolving needs of pharmaceutical and biotech customers
Quality Control Capabilities and Instrumentation
- Control of trace organic impurities in solvents to the part per billion (PPB) level
- UPLC-PDA/MS (Ultra-Performance Liquid Chromatography)
- GC-FID, GC-ECD
- UV spectroscopy
- Fluorescence spectrometry
- Karl Fischer titration