TrueForm Quality. Comfort. Dexterity.
TrueForm™ is made for those who rely on daily hand protection to provide ease of use and high dexterity. With TrueForm, quality is designed into every single style using exacting manufacturing processes for tip-to-cuff consistency, glove after glove.
Product Selection
TrueForm TF-036-095-LB
One of the thinnest single-use nitrile exam gloves available, providing super fine dexterity and tactile sensitivity.
TrueForm TF-050-095-NRG-LG
Single-use nitrile gloves with EnerGel™ technology, hydrate and sooth dry, chapped skin. Gloves are 100% air inspected and consistent thickness tip to cuff.
TrueForm TF-050-095-RB
Single-use nitrile exam gloves. The new lighter standard for everyday exam use. Consistent quality glove to glove.
TrueForm TF-051-095-PHN-BL
Nitrile single-use exam glove with pH Natural® helps maintain pH levels for improved hand health.
TrueForm TF-060-095-BK
Black single-use nitrile gloves are 100% air inspected and consistent thickness tip to cuff.
TrueForm TF-066-095-SB
Premium nitrile single-use exam gloves at 6.6 gram weight and 0.65 AQL. Gloves are 100% air inspected and consistent thickness tip to cuff.
TrueForm TF-075-120-LG
Extended-cuff single-use nitrile exam gloves. Providing extra protection and tested for use with harmful chemotherapy drugs.
TrueForm TF-090-095-TTK-NT
TracTek® performance grip technology latex glove with a high density pattern for grip in wet/oily conditions.