MegaMan Tougher. Rougher. Ready.
MegaMan® pushes the boundaries of single-use hand protection performance – with new levels of abrasion resistance, sweat management, and comfort that deliver in exceedingly harsh environments. When protection performance matters most, it’s no wonder MegaMan gets the call for the toughest jobs.
Product Selection
MegaMan MM-128-110-DRK-BK
Heavy duty nitrile gloves with DriTek® sweat absorption lining. Multilayers last 3X longer with ANSI Level 3 abrasion resistance.
MegaMan MM-128-110-DRK/NRG-HVGR
Hi-Vis green heavy duty nitrile gloves with DriTek® sweat absorption lining and EnerGel™ hand rejuvenating technology last 3X longer.
MegaMan MM-211-120-BL
Heavy duty nitrile gloves with extended cuff for added protection. Gloves are 100% air inspected and consistent thickness tip to cuff.