TrueForm® TF-090-095-TTK-NT TracTek® Performance Grip for Latex Gloves, SW Safety Solutions

Supplier: Avantor
76051-012BX 46.58 CAD
76051-012 76051-016
TrueForm® TF-090-095-TTK-NT TracTek® Performance Grip for Latex Gloves, SW Safety Solutions
Gloves
Nothing gets the slip on wearers when using these high-performance grip gloves. Designed to provide barrier protection against chemicals, pathogens, and more when wearers need to maximize dexterity.

  • TracTek® Grip Technology is engineered to maximize surface contact and channel away liquids
  • High tactile sensitivity and comfort of natural latex
  • Natural latex provides advanced chemical protection
  • Helps prevent skin irritation and sample contamination
  • Optimal thickness for durability and tactile sensitivity

These gloves are tear-resistant beaded cuffMore Product Information
Order Now

SPECIFICATIONS

Model TF-090-095-TTK-NT
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Disposable / reusable Disposable
Latex/Latex-free Latex
Powdered/Powder-free Powder-free
Finish Finger Textured
Cuff style Beaded
Cuff length 242 mm (9.5")
Cuff thickness 5.1 mil (0.10 mm)
Finger thickness 7.9 mil (0.17 mm)
Palm thickness 6.3 mil (0.14 mm)
FDA regulated +
Tensile strength 22 MPa
Ultimate elongation 670%
EN norm EN 420:2003; EN 455:2000-2015
Food contact approved +
ASTM No. D3578; F1671-97; F739-12

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR