Inlet Septa for Gas Chromatography
Advanced Green inlet septa eliminate sticking and reduce coring. Bleed temperature optimized (BTO) inlet septa are preconditioned and ready to use with low-bleed, mass spec capillary columns. Our long-life septa are specially formed for extended use, meaning less downtime. Explore Agilent's complete portfolio of gas chromatography septa to ensure your GC is operating at peak performance.
Advanced Inlet Septa
With our premium non-stick septa, you'll never have to scrape bits of septum out of your inlet.
Bleed Temperature Optimized (BTO) Septa
Bleed temperature optimized septa are specifically designed for operation at temperatures up to 400°C without bleeding into the sample flow path.
General Purpose Septa
Agilent’s general-purpose septa are made from an enhanced injection-molded silicone rubber material.
Long Life Septa
With Agilent's premium non-stick septa, you'll never have to scrape bits of septum out of your inlet.
Merlin Microseal
The Merlin Microseal is a microvalve alternative to the conventional silicone rubber septa used in gas chromatography (GC).