Capillary Tubs and Accessories
Agilent provides all the necessary gas chromatography accessories and tooling for you to perform maintenance on your capillary columns. Our extensive portfolio includes column hangers and baskets, capillary rinse kits, installation kits, and preswaging tools.
Capillary Column Installation Kit
The capillary column installation kit provides everything needed to easily install a GC capillary column.
Capillary Column Rinse Kit
This kit makes solvent rinsing easy! Solvent rinsing removes many contaminants that accumulate in columns.
Column Baskets & Hangers
Agilent offers a range of column baskets and hangers
Column Installation Pre-Swaging Tools
Optimize GC performance and reproducibility by ensuring a proper length of column into the inlet, time after time.
Fused Silica Tubing Cutters
Tool and ceramic wafer that allow you to make clean and easy cuts in fused silica, glass, and aluminum-clad capillary columns in GC systems.