Microbiology
For premium quality and performance in quality testing labs, you can count on Corning. In our continuous efforts to improve efficiencies and develop new tools for microbiology applications, we have scientists working in Corning labs doing what you do every day, across the globe. From sampling and preparation to bacteria growth and analysis, our technical experts understand your challenges and the changing regulatory requirements that shape your day-to-day processes.
Corning® Media Bottles, PET, with IATA-Validated Screw Cap
Corning® PET media bottles are equipped with a screw cap designed to ensure leak-proof performance.
- Available in four sizes
- Packaged in convenient, shrink-wrapped trays within an outer bag to ensure cleanliness
- Break-resistant bottles are an ergonomic and easy-to-handle alternative to glass
- Molded graduations for accurate measurements
- Innovative HDPE screw cap meets IATA requirements to ensure leak-proof air transport conditions
These bottles with IATA-validated screw caps are designed for safe, secure storage of tissue culture media, sera, buffers, and other aqueous solutions. The PET resin makes the bottle lightweight, break-resistant, and as transparent as glass. The screw cap is designed to ensure leak-proof performance and meets International Air Transport Association (IATA) requirements for primary packaging used in air transportation.
Certifications: Bottles have Sterility Assurance Level (SAL) of 10⁻⁶. Bottles are validated USP Class VI, noncytotoxic, nonhemolytic, and nonpyrogenic.
Sampling
Preparation and Testing
Analysis