Chemistry
Corning has been a valued supplier to chemists for nearly a century since the introduction of PYREX® low expansion borosilicate glass that is the quality standard for labware because of its optical clarity, strength, heat, and chemical resistance.
PYREX® Distilling Flasks, Corning
PYREX® borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Break-resistant bottles are an ergonomic and easy-to-handle alternative to glass
The sidearm of the 125 ml flask is located approximately 137 mm from the bottom of the flask as required in ASTM E1405 and can be used in ASTM Tests D86 and D447. This size has a heavy-duty neck.
The sidearm on the 250 ml size is placed 77 mm below the top of the neck at a 75° angle and can be used with thermometers calibrated for 76 mm immersion.
Separation/Purification
Extraction/Concentration
Weighing/Mixing/Heating