Magnetic 3D Cell Culturing
Magnetic 3D Cell Culturing 3D in a fast and easy 2D workflow
The core technology is the magnetization of cells with NanoShuttle™-PL. The cells can be aggregated with magnetic forces, either by levitation or printing, to form structurally and biologically representative 3D models in vitro.
- The advantages of magnetic cell culture include
- Mimicking native tissue environment
- Rapid 3D model formation within hours
- No specialized equipment, media, or artificial substrate
- Easy to handle / no sample loss
- Allows co-culture
CELLSTAR® Cell Culture Vessels with Cell-Repellent Surface
In contrast to standard tissue culture surfaces which are optimized to enhance conditions for cell attachment, the cell-repellent surface has been developed to effectively prevent cell attachment. CELLSTAR® cell culture vessels with a cell-repellent surface reliably prevent cell attachment in suspension cultures of semi adherent and adherent cell lines where standard hydrophobic surfaces generally used for suspension culture are insufficient.
Magnetic 3D Cell Culture Products
VWR PN | GBO PN | Product Descriptions |
76324-742 | 657840 | 6-Well Bio-Assembler Kit |
76324-760 | 662840 | 24-Well Bio-Assembler™ Kit |
76324-014 | 655840 | 96-Well Bioprinting Kit, Clear Plates |
76324-016 | 655841 | 96-Well Bioprinting Kit, Black µClear Plates |
76324-018 | 655846 | 96-Well BiO Assay™ Kit |
76324-020 | 655849 | 96-Well Bio Assay Kit™ & Imaging System |
76324-768 | 781840 | 384-Well Bioprinting Kit, Clear Plates |
76324-770 | 781841 | 384-Well Bioprinting Kit, Black µClear Plates |
76324-772 | 781846 | 384-Well BiO Assay™ Kit |
76324-282 | 781849 | 384-Well BiO Assay™ Kit & Imaging System |
76337-248 | 781850 | 384-Well Ring Drive |
76337-250 | 789830 | 1536 well Spheroid Drive |
76337-252 | 789837 | 1536 WELL HOLDING DRIVE |
76324-744 | 657841 | NanoShuttle™-PL Refill, 600 µL Vial (1) |
76324-746 | 657843 | NanoShuttle™-PL Refill 3 Pack, 600 µL Vials (3) |
76324-748 | 657846 | NanoShuttle™-PL Refill 6 Pack, 600 µL Vials (6) |
76324-750 | 657847 | NanoShuttle™-PL Refill 6 Pack with FREE iPod™ (Only with purchase of BIO Assay™ 655849 or 781849) |
76324-754 | 657852 | NanoShuttle™-PL Refill 12 pack, 600 µl Vials |
76324-752 | 657850 | MagPen™ 3 Pack |
76324-024 | 657860 | Imaging Kit |