Greiner Bio-One Separation
With Leucosep™, Greiner Bio-One optimized density gradient centrifugation whilst making it user-friendly. Alongside this, OncoQuick® was developed to extend the spectrum of applications to deal specifically with oncological targets.
Leucosep™ Centrifuge Tubes
Leucosep™ Tubes 12 mL and 50 mL sizes. Designed for easy separation of human monocytes and lymphocytes from peripheral blood.
- Easy to fill without cumbersome layering steps
- Easy recovery of interphase
- No re-contamination with unwanted red blood cells
- PBMC isolation from whole blood in 15 min
- Un-filled or pre-filled with Leucosep™ separation medium
Learn More about Greiner Bio-One products
Product Selection
EASYstrainer™ Cell Strainer
EASYstrainer™ cell strainers are filters for Cell suspensions and primary cell isolates. Sterile. Fits all standard 50ml tubes. Venting slot for fast filtration
Leucosep™ Centrifuge Tubes
Leucosep™ developed for optimal separation of lymphocytes and peripheral mononuclear cells from human whole blood and bone marrow. Key feature is the porous barrier incorporated in the centrifuge tube
OncoQuick
OncoQuick® is a simple-to-use, rapid and efficient system for the enrichment of circulating tumor cells that are released into the blood by a solid epithelial tumor or malignant melanoma...