Diagnostic Tests—Adjuvant Complete to Neisseria meningitidis Antiserum, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
223091 223101 227901 231141 240658 221591 222361 222371 222501 223021 223142 227891 227911 227921 229101 229701 231131 240661 263810 263910 264010 225851
CA90001-632EA 369.08 CAD
CA90001-632 CA90001-732 CA90001-736 CA90001-744 CA90001-950 CA90002-208 CA90003-124 CA90001-390 CA90001-628 CA90001-630 CA90001-636 CA90001-640 CA90001-644 CA90001-646 CA90001-648 CA90001-652 CA90001-656 CA90001-718 CA90001-742 CA90001-948 CA90001-952 CA90001-956 CA90002-002 CA90002-004 CA90002-010 CA90002-020 CA90002-040 CA90002-206 CA90003-126 CA90003-746 CA90003-748 CA90010-128 CA90010-142
Diagnostic Tests—Adjuvant Complete to Neisseria meningitidis Antiserum, BD Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
Antigens and antisera used for slide agglutination tests and immunofluorescence. BD Difco Listeria O Antigens Types 1 and 4 (Slide) are suspensions of appropriate serotypes containing 0.3% formaldehyde as a preservative
Store at 2 to 8 °C (35.6 to 46.4 °F).