SeaKem® Agarose, Lonza
Supplier: Lonza
50000 50001 50004 50070 50071 50074 50150 50152 50002 50005
CA12001-866EA 501.44 CAD
CA12001-866 CA12001-868 CA12001-870 CA12001-892 CA12001-894 CA12001-896 CA12001-910 CA12001-912 CA12002-102 CA12002-104
SeaKem® Agarose, Lonza
Electrophoresis Gels Agarose and Acrylamide
SeaKem® is a high gel strength, low EEO, standard gelling temperature agarose perfect for electrophoresis and other molecular biology applications.
  • All Lonza agarose is quality tested to certify performance
  • High gel strength
  • Low EEO
  • Standard melting temperature
Agarose is a key ingredient in your research
Lonza’s SeaKem® LE was the first and is still the world’s most trusted agarose for nucleic acid electrophoresis. Ideal for routine analysis of nucleic acids by gel electrophoresis and blotting, each SeaKem LE gel sharply resolves DNA and providesMore Product Information