Glutathione peroxidase activity kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-158
89141-230EA 1220 CAD
89141-230
Glutathione peroxidase activity kit, Enzo Life Sciences
Assays Enzyme Assays
For the measurement of glutathione peroxidase in cell lysates, plasma, and tissue from any species.
The Glutathione Peroxidase activity kit is a convenient 96-well plate based assay for the higher throughput analysis of glutathione peroxidase activity in a variety of sample matrices in just 30 minutes.
Ordering information: Contents: Microtiter plate, Assay buffer concentrate, Reaction mix, Glutathione reductase, GSSG