Masterflex® Solenoid-Operated Pinch Valve For Masterflex® Pump Tubing, Avantor®
Supplier: CLARK SOLUTIONS
PK-1605NC-24V PK-1605NO-12V PK-4805NC-12V PK-4805NC-24V PK-4805NO-24V PK-6405NC-24V PK-1605NC-12V PK-3205NC-24V PK-3205NO-12V PK-3205NO-24V PK-4805NO-12V PK-6405NC-12V PK-6405NO-12V PK-6405NO-24V
MFLX98306-02EA 237 CAD
MFLX98306-02 MFLX98306-04 MFLX98306-08 MFLX98306-16 MFLX98306-18 MFLX98306-22 MFLX98306-26 MFLX98306-00 MFLX98306-10 MFLX98306-12 MFLX98306-14 MFLX98306-20 MFLX98306-24 MFLX98306-28 MFLX98306-30
Masterflex® Solenoid-Operated Pinch Valve For Masterflex® Pump Tubing, Avantor®
Valves Solenoid Valves
Specifically designed for Masterflex® pump tubing.
  • Use with tubing that has durometer rating of 50 or less such as C-Flex® or silicone—7.25 psi max
  • Excellent for clean processes—fluid stays contained within tubing
  • Use for on/off flow control applications
Easily control the flow through your Masterflex tubing.