Masterflex® Solenoid Pinch Valve, Normally-Open, for L/S 25 tubing; 24 VDC
Supplier: CLARK SOLUTIONS
VWR Item#MFLX98306-22
Your Organization's Price:
Retrieving
Specifically designed for Masterflex® pump tubing.
  • Use with tubing that has durometer rating of 50 or less such as C-Flex® or silicone—7.25 psi max
  • Excellent for clean processes—fluid stays contained within tubing
  • Use for on/off flow control applications
Easily control the flow through your Masterflex tubing.