Masterflex® Solenoid Pinch Valve, Normally-Closed, for L/S 25 tubing; 24 VDC
Supplier: CLARK SOLUTIONS
Specifically designed for Masterflex® pump tubing.
- Use with tubing that has durometer rating of 50 or less such as C-Flex® or silicone—7.25 psi max
- Excellent for clean processes—fluid stays contained within tubing
- Use for on/off flow control applications
Easily control the flow through your Masterflex tubing.