Macro-Vue™ RPR Circle Card Test Kits, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
275539 274449 275005
CA90003-960EA 226.94 CAD
CA90003-960 CA90003-924 CA90003-942L
Macro-Vue™ RPR Circle Card Test Kits, BD Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
These rapid plasma reagin (RPR) test kits are developed for use in large-scale testing in public health and clinical laboratories
Kits use mechanical rotation, antigen suspension, and specially prepared, plastic-coated test cards for the serologic detection of syphilis. The recommended speed for rotation is 100rpm. The rotator should circumscribe a circle approximately 2cm in diameter in the horizontal plane. Refrigeration is recommended for storing the antigen suspension ampules. The disposable BD Dispenstirs™ devices allow the transfer of specimen to the card surface. An antigen dispensing bottle with a needle serves for accurate drop delivery of antigen. Spot cards feature circle spots 18mm in diameter.
Kit contents:
3 mL Antigen
Ampules
10-Spot Cards
Capillaries, 0.05 mL
Antigen Dispensing Bottle Stirrers
20-Gauge Dispensing Needle
Order Now
SPECIFICATIONS
Environmentally Preferable