Matrix Metalloproteinase-2 (MMP-2) Colorimetric Drug Discovery Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK408-0001
89156-466EA 1063.34 CAD
89156-466
Matrix Metalloproteinase-2 (MMP-2) Colorimetric Drug Discovery Kit, Enzo Life Sciences
Assays Enzyme Assays
A QuantiZymeTM Assay System
The MMP-2 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-2 (MMP-2, gelatinase A) using a thiopeptide as a chromogenic substrate. The assays are performed in a convenient 96-well microplate format. The kit is useful to screen inhibitors of MMP-2, a potential therapeutic target.
Delivery information: Kit contains MMP-2 enzyme (human) (recombinant), substrate, inhibitor, assay buffer, 96-well microplate.