The Ideal Workstation for Sterility Testing
Create an ISO 5 / Grade A equivalent workspace nearly anywhere in your facility with the Bioquell Qube isolator. Host the Bioquell Qube outside of a Grade A environment to save space in the cleanroom, reduce operating costs, require less gowning, and more. The system offers a closed, validated, bio-decontaminated environment, allowing you to work comfortably and efficiently.
The Bioquell Qube brings efficiencies and reliability to your sterility testing process. Available in two, four, or six glove configurations, with up to two material transfer hatches, the Bioquell Qube’s modular design can meet your company’s sterility testing Needs. The process is simple, effective, and efficient thanks to Bioquell’s hydrogen peroxide vapor technology and Bioquell Hydrogen Peroxide Sterilant.*
Start with the simple to navigate touch screen to begin a validated decontamination cycle that reaches throughout every chamber. Close the latch between the chamber containing the vaporizer and the adjacent workstation hosting the sterility pump. You now have an aseptic environment to perform your testing that can last at least seven days before requiring repeat decontamination.
Place your testing supplies on the racking in the vaporizer’s chamber. When a cycle is started in the chamber with the supplies, exposed surfaces receive a validated 6-log sporicidal kill in as little as twenty minutes.
The decontamination supplies are transferred to the workstation with the pump to begin testing. Operators can now safely perform tests while additional supplies are being loaded and decontaminated in the adjacent workstation, creating a nearly unimpeded workflow.
- Integrated with Ecolab Bioquell hydrogen peroxide vapor technology
- Validated cycles create 6-log sporicidal kill on exposed surfaces
- Utilizes EPA registered sterilant 35% w/w aqueous hydrogen peroxide solution
- Creates validated Grade A / ISO 5 environment
- Customize to meet your working needs
- Configurations up to 6 gloves, 2 material pass-throughs or rapid transfer ports
- One inch tri-clover port(s) in any chamber
- Rapid transfer port options for aseptic transfer and more
- Select an optional integrated sterility test pump from Merck Millipore or host your own benchtop pump
- four levels of viable and non-viable environmental monitoring to select to ensure strict quality standards are met
- racking solutions allow for high throughput and provide operators with ample room to work comfortably
- ergonomic features allow operators to sit and work comfortably at their station for extended periods, includes angled glass, adjustable lighting intensity, and wide glove ports providing excellent range of motion
- Full traceability and quality control through RFID tags on hydrogen peroxide bottle and printed cycle reports
- Optional logging capability with 21 CFR part 11 compliant software
- From order to installation and validation, the Bioquell Qube can be ready for use in as little as 16 weeks
- Does not need to be ducted into HVAC
- Operates in negative or positive pressure
- Fits through most standard doorways, and requires only a standard outlet to function.
- Bio-decontaminate incoming materials while working in another chamber
- No construction or electrical work required for installation
- Supports GMP compliance
- Bioquell Hydrogen Peroxide Sterilant is an EPA Registered Sterilant (EPA Registration Number: 72372-1-86703)
Bioquell Qube, a modular isolator with integrated Bioquell bio-decontamination technology
Create an aseptic workspace with Ecolab’s Bioquell Qube, a configurable isolator integrated with Bioquell hydrogen peroxide vapor technology.