Controlled Environments
Products including Tyvek® for use in critical environments within the biotechnology, pharmaceutical, microelectronics, semiconductor, food and beverage, medical device, and other industries.
Sterile Cleanroom Manufacturing
Tyvek® IsoClean® Garments and Tyvek® Micro-Clean® 2-1-2 Garments
- Comfortable, lightweight and durable
- Individually packaged garments offered as sterile or clean-processed & sterile
- Garments available gamma sterilized to an SAL of 10-6 with full traceability on all sterile apparel
- Serged or bound seam styles are available
Non-Sterile Cleanroom Manufacturing
Tyvek® IsoClean®, Tyvek® Micro-Clean® 2-1-2, ProClean® and ProShield® Garments
- Delivers the particle barrier, durability and comfort in use
- Available as individually packaged and clean processed as well as bulk packaged
- Variety of fabrics to meet your specific applications and needs
Controlled Environment and General Protection
Tyvek® IsoClean®, ProClean® and ProShield® Garments
- Delivers the particle barrier, durability and comfort in use
- Offers protection against non-hazardous particulates and light liquid splash
- Lightweight and low linting