Controlled Environment and General Protection
DuPont controlled environment garments offer comfortable, lightweight and durable protection for your manufacturing environment. Serged and bound seam garments are available in a variety of materials including DuPont™ Tyvek, DuPont® ProShield® SMS and DuPont® ProShield® microporous film.
Tyvek® IsoClean® bulk packaged garments
- Comfortable, lightweight and durable
- Individually packaged garments offered as sterile or clean-processed & sterile
- Garments available gamma sterilized to an SAL of 10-6 with full traceability on all sterile apparel
- Serged or bound seam styles are available
ProShield® 50 and ProShield® 60
- Made from a microporous composite fabric
- Offers protection against non-hazardous particulates and light liquid splash
- Lightweight and low linting
- Two basis weights, 50 gsm and 60 gsm
ProShield® 10
- SMS garments are optimized for comfort, softness and breathability
- Available in a variety of coverall styles in a choice of white, blue and gray
- Designed for a range of cleanroom manufacturing and controlled environment applications.
- Nonhazardous dry particle and light liquid splash applications
DuPont™ ProShield® 30 and Dura-Trac™ Shoe & Boot Covers
- Made from spun bonded polypropylene with a polyethylene coating
- Provides exceptional skid resistance
- May be suitable for Cleanroom and General Maintenance jobs
DuPont™ Sierra™ Masks
- Provides balance of bacteria and particle filtration efficiency and breathability