Liners for Contamination Control
Micronova Bags and Liners are used for a variety of containment functions in the cleanroom and aseptic areas. The Liners, available sterile, are used for storage of raw materials, disinfectant containment, disposal, and segregation of biohazardous materials.
BucketLiners™
An impermeable barrier made from LDPE/Nylon laminate is ideal for lining buckets containing liquid solutions or to line totes for transporting product parts or raw materials. Excellent for liquid containment and material transport.
BinLiner™ Cleanroom Bags
These LDPE liners are Ideal for a variety of uses ranging from the collection & disposal of waste to the transport of parts or raw materials in cleanrooms & aseptic environments. Available gamma irradiated or non-irraidated in multiple thicknesses and cap
BioHazard and Hazard BinLiner™ Bags
These LDPE liners are Ideal for a variety of uses ranging from the collection and disposal of waste to the transport of parts or raw materials in cleanrooms and aseptic environments. Available gamma irradiated or non-irriadated in multiple thicknesses an
Stainless Steel BinLiner™ Stands
The perfect complement to the BinLiner™ Bags, with easy-to-clean plastic corners and stainless steel supports that do not touch the floor. Ideal for cleanroom material disposal, garment collection, and raw material or product storage. Multiple sizes avail