Rheo Flexibles: Single-Use Containment and Transfer Technology
Rheo Flexibles is a division of Rheo Engineering that provides a full range of single-use containment and powder transfer products for the life sciences industry. Rheo Flexibles products are made from high quality film that is fully compliant with all of the regulatory requirements of the pharmaceutical industry. All Rheo Flexibles products are produced and packaged in an ISO 8 cleanroom and come with a complete batch tracing documentation package. Rheo offers many standard products that are able to ship quickly, and we offer custom solutions as well.
RheoPacs (Single-Use Bags)
The RheoPac family of products offer an excellent alternative to stainless intermediate transfer containers.
Transfer Isolators
The Rheo Transfer Isolator is designed to allow operators to safely remove product from containers and transfer them into the next step of the process. Single-Use Isolators offer a quick deployment containment solution with no cleaning costs associated with stainless isolator systems.
Continuous Liners
Rheo Continuous Liners are an excellent packoff solution for transferring materials from a process into transport containers while maintaining a high level of containment.
RheoCrimp
The RheoCrimp is the latest in easy-to-use cut and crimp separation technology. These crimps are very easy to use as they offer a wide opening range for capturing bulky liner material. The RheoCrimp Kit includes the tools needed for this crimp and cut process as well as a pack of 50 crimps.
Rheo Flexibles Video
Rheo Flexibles is a division of Rheo Engineering that provides a full range of single-use products for the safe handling and transfer of high value and potentially hazardous powders.
Custom Solutions
Rheo Flexibles offers custom single-use containment and transfer systems. Partner with our inside engineering team to develop the right solution for your process.