Azure cSeries Advanced Imaging Systems
The c300-c600 offer imaging capabilities for chemiluminescence, UV, RGB, NIR, blue light, and white light, depending on the model that fits your needs. The c600 offers the most flexible imaging options, but each model can be upgraded to suit your lab’s needs as they change.
c600 | c500 | c400 | c300 |
---|---|---|---|
Ultimate Western SystemIR Fluorescence |
Infrared Imaging SystemIR Fluorescence Upgradeable to c600 |
Visible Fluorescent |
Darkroom EliminatorChemiluminescence Upgradeable to c600, c500, and c400 |
![]() |
![]() |
![]() |
![]() |
cSeries Specifications | c600 | c500 | c400 | c300 |
---|---|---|---|---|
Camera | 8.3 MP | |||
Cooling | -50°C regulated cooling | |||
7 Position Filter Wheel | ✓ | ✓ | ✓ | ✓ |
UV 302/365 nm | ✓ | ✓ | ✓ | ✓ |
EPI Blue LED | ✓ | ✓ | ✓ | ✓ |
Chemi | ✓ | ✓ | ✓ | ✓ |
Visible Fluorescent Westerns | ✓ | ✓ | ||
NIR Westerns | ✓ | ✓ | ||
Field of View | 20 × 15 cm | |||
Footprint (W × H × D) | 38 × 55 × 36 cm |