Protector® Fume Hood Airflow Monitors, Service Fixture Kits, and Duplex Receptacle Kits, Labconco
Supplier: Labconco
9413300 9413400 985340010814 985770010814 9410300 9808300 9808400 9808500 9808700 9808800 9808900 9809100 9809200 9809300 9809700 9827900 9828000 9851100 9851500 985420010814
89260-082EA 2180.4 CAD
89260-082 89260-084 89260-110 89260-112 89260-786 89260-086 89260-088 89260-090 89260-092 89260-094 89260-096 89260-098 89260-100 89260-102 89260-104 89260-106 89260-108 89260-114 89260-116 89260-118
Protector® Fume Hood Airflow Monitors, Service Fixture Kits, and Duplex Receptacle Kits, Labconco
Fume Hoods and Enclosures Chemical Fume Hoods
There are many accessories available for customizing Protector® Laboratory Hoods including Service Fixture Kits, Duplex Receptacle Kits and Guardian™ Airflow Monitors.
Each Service Fixture Kit includes one remotely-controlled service fixture or gooseneck with valve and outlet tubing, color-coded fixture knob and color-coded hose connector. Choose from a variety of configurations. All kits are compatible with Protector® Premier®, XStream®, PVC and XL™ Hoods. Gooseneck kits are also compatible with Protector® Stainless Steel Perchloric Acid Hoods.
Duplex Receptacle Kits include wiring. Guardian™ Airflow Monitors alert operators to low airflow conditions. Guardian™ Digital Airflow Monitors provide a digital display of face velocity as well as an audible alarm.
Ordering information: Base cabinets, blowers and work surfaces are sold separately.