Potassium iodide 15% w/v aqueous solution Stabilised
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Dipotassium diiodide, Kali iodide
R6305000-500B
R6305000-1B
RC630516EA
202.24
CAD
RC630516
RC630532
Potassium iodide 15% w/v aqueous solution Stabilised
Potassium iodide
Formula:
Kl MW: 166 g/mol Boiling Pt: 1330 Melting Pt: 686 Density: 1.1086 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011405 CAS Number: 7681-11-0 Merck Index: 13,07727 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 999) | 15.0 ± 0.1 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...