Potassium iodide 2% (w/v) in aqueous solution

Supplier: Ricca Chemical
Danger

R6288000-500B R6288000-1B
RC628816EA 86.9 CAD
RC628816 RC628832
Potassium iodide 2% (w/v) in aqueous solution
Potassium iodide
Formula: Kl
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0149 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 2.00 ± 0.10 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR