Potassium iodide ≥99%, granules, BAKER ANALYZED® ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Dipotassium diiodide, Kali iodide
BAKR3162-05
BAKR3162-07
BAKR3162-09
CAJT3162-1CS
3282.37
CAD
CAJT3162-1
CAJT3162-5
CAJT3162-07
CAJT3162-9
CAJT3162-11
Potassium iodide ≥99%, granules, BAKER ANALYZED® ACS
Potassium iodide
Potassium Iodide in granular form.
ACS Reagent.
Formula:
Kl Boiling Pt: 1330 °C (1013 hPa) Melting Pt: 686 °C Density: 3.10 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011405 CAS Number: 7681-11-0 EINECS: 231-659-4 Merck Index: 12,07809 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (KI) | ≥99.0 % |
Insoluble Matter | ≤0.005 % |
Loss on Drying at 150°C | ≤0.2 % |
pH of 5% Solution at 25°C | 6.0 - 9.2 |
Chloride and Bromide (as Cl) | ≤0.01 % |
Phosphate (PO₄) | ≤0.001 % |
Sulfate (SO₄) | ≤0.005 % |
Barium (Ba) | ≤0.002 % |
Calcium (Ca) | ≤0.002 % |
Magnesium (Mg) | ≤0.001 % |
Sodium (Na) | ≤0.005 % |
Iodate (IO₃) | ≤3 ppm |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤2.0 ppm |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...