Potassium iodide 5% (w/v) in aqueous solution stabilised analytical reagent

Supplier: Ricca Chemical

Synonyms: Dipotassium diiodide, Kali iodide

R6290000-500B R6290000-4B
RC629016EA 97.96 CAD
RC629016 RC6290-1
Potassium iodide 5% (w/v) in aqueous solution stabilised analytical reagent
Potassium iodide
Formula: Kl
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 5.00 ± 0.10 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR