Potassium iodide 1.00 N in aqueous solution

Supplier: Ricca Chemical

Synonyms: Dipotassium diiodide, Kali iodide

R6318000-1C R6318000-4C R6318000-500C R6318000-20F
RC6318-32EA 652.54 CAD
RC6318-32 RC6318-1 RC6318-16 RCR6318000-20F
Potassium iodide 1.00 N in aqueous solution
Potassium iodide
Formula: Kl
MW: 166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay (vs. Silver Nitrate/Eosin Y) at 20 °C (traceable to NIST SRM 999) 1.00 ± 0.01 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR