Potassium iodide 10% (w/v) in aqueous solution stabilised analytical reagent

Supplier: Ricca Chemical

Synonyms: Dipotassium diiodide, Kali iodide

R6300000-500B R6300000-1B R6300000-20F R6300000-4B
RC630016EA 129.56 CAD
RC630016 RC630032 RC63005 RC63001
Potassium iodide 10% (w/v) in aqueous solution stabilised analytical reagent
Potassium iodide
Formula: Kl
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 10.0 ± 0.1 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR