Ward's® Live Vorticella Culture
Supplier: Ward's Science
VORT30/35
470176-588EA 15.7 CAD
470176-588 470176-590 470180-176
Ward's® Live Vorticella Culture
Specimens Live Specimens Protists - Living Material
This organism is easily recognized by its trumpet-shape, and can commonly be found in freshwater ponds and streams. It reproduces by budding.
  • Compare to Stentor for differences in cilia arrangement
  • Reproduces both sexually (via conjugation) and asexually (via budding)
  • Vital-Staining is available for this culture
  • Cytoplasmic elements are stained blue on your live culture, to allow for better visibility
  • Skip the wet-mount
  • Available as a set of 5 Readyslides
  • Tapered end of the container fits directly on your microscope
Vorticella is commonly mistakenMore Product Information