Ward's® Streak and Color Collection
Supplier: Avantor
Ward's®
WARD470025-252
470025-252KT 55.39 CAD
470025-252
Ward's® Streak and Color Collection
Specimens Mineral Specimens Mineral Collections
Clear examples of a key method of identification.
  • Housed in a compartmented collection box
  • Nine 1×2" numbered samples
  • Identification list included
  • Streak plate included
The powder a mineral produces is at times a different color from the mineral itself. Samples in this set were chosen for dramatic color and streak. Minerals: Chromite, galena, hematite, limonite, microcline, pyrite, rose quartz, sphalerite, sulfur.