Ward's® Live Spirogyra Culture
Supplier: Avantor
470176-596EA 48.9 CAD
470176-596 470194-676
Ward's® Live Spirogyra Culture
Specimens Live Specimens Protists - Living Material
Spirogyra is a genus of filamentous green algae with chloroplasts arranged in a helical (spiral) pattern.
  • Excellent food source for freshwater snails and fish
  • Undergoes both sexual and asexual reproduction
Spirogyra are a genus of Chlorophyte algae that can be found in freshwater lakes and ponds. They are characterized by green plastids and cell walls composed of cellulose.
Ordering information: Genus: Spirogyra. Species: not specified.