Ward's® Pyroxene Thin Section Set
Supplier: VWR
WARD470024-860
470024-860KT 971.79 CAD
470024-860
Ward's® Pyroxene Thin Section Set
Specimens Mineral Specimens Mineral Collections
Great value for the microscopic study of Pyroxene minerals
  • Specially Selected by Wards® staff Geologists
  • 12 Thin Sections
  • Includes Key for Easy Identification
Specially selected thin sections of important rock forming minerals. Set may contain more than one example of the same mineral to depict variations in occurrence and distinct mineral associations.
Slides (12): acmite augite (2) diopside, enstatite, fassaite, hedenbergite, hypersthene, jadeite, johannsenite, pigeonite, spodumene