Ward's® Live Paramecium multimicronucleatum Culture
Supplier: Avantor
470176-726EA 44.15 CAD
470176-726 470199-682
Ward's® Live Paramecium multimicronucleatum Culture
Specimens Live Specimens Protists - Living Material
Paramecium multimicronucleatumis one of the largest species of Paramecium. This ciliate contains a single macronucleus and four or more micronuclei. Grows in Dilute Alfalfa medium (Hay Medium).
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Delivery information: Please specify a delivery date during checkout.