1.5 mw Helium-Neon Modulated Laser
Supplier: INDUSTRIAL FIBER OPTICS INC SE
IFHN15M
470111-380EA 1548.4 CAD
470111-380
1.5 mw Helium-Neon Modulated Laser
Laser Pointers
Ideal Classroom Instrument for all Optical Work
  • Class: IIIa
  • Case Size: 14-1/2 x 3-3/4 x 2-1/4"
Ruggedly constructed lasers with hard-seal tubes for long life. Each is housed in a transparent blue, impact-resistant acrylic housing with positive grip rubber feet. The output end incorporates a manual beam-blocking safety shutter, and has a threaded mount for standard beam-shaping accessories. The rear panel carries the power switch and jack for connecting the included power adapter (UL approved, for 115V AC). A standard 1/4-20 threaded hole in the base accepts a stand rod for optical bench or tripod mounting (not included on 5.0mW model). The lasers emit red light with a wavelength of 632.8nm in the TEM00 mode. Includes full color instruction manual. Continuous Wave (CW) Lasers emit a continuous beam of light with only small output power variations after warm-up. They are suitable for most applications. Modulated Lasers have additional jacks (3.5mm audio) to allow a varying signal of 600Hz–20kHz to be input for modulating the output beam intensity. This is used for measuring the speed of light and for optical signal transmission experiments. Both will accept modulation from audio, video or signal generator sources to demonstrate laser communication or be used to measure the speed of light.