Classroom Collection of Minerals and Rocks
Supplier: SCOTT RESOURCES 2, LLC SE
SB46077
470112-326KT 128.25 CAD
470112-326
Classroom Collection of Minerals and Rocks
Specimens Mineral Specimens Mineral Collections
Classroom Collection of Rocks and Minerals Designed for General Classroom Use
  • Great for Classroom Use
  • Study Guide Included
  • Covers all three rock types and minerals
For basic study of mineralogy, this collection contains 50 rock and mineral samples coming largely from the Western region of the United States. Specimens are grouped into the following categories for study:
Rock-forming minerals; igneous rocks; metamorphic rocks; sedimentary rocks; ores of common metals and non-metals; gemstones and scale of hardness samples.
Study guide is included. Sample sizes: 1" x 1".