ACT + Fast Anti Choke Training Vests
Supplier: American 3B Scientific
1017938 1014589
470125-138EA 352.65 CAD
470125-138 470125-140
ACT + Fast Anti Choke Training Vests
Models Human Biology Models
Teach The Heimlich Maneuver
  • Available with, or without "back slap"
  • Practice on a manikin or individual
An invaluable tool for instructors to safely teach students to administer the Heimlich maneuver on a live partner. This strap-on vest makes CPR education more realistic and fun. When performed correctly, a foam plug shoots into the air to simulate the release of the barrier that is lodged in the airway. The Red version also includes a Back Slap Pad for practicing ARC and ERC protocols. The trainers can be used standing, sitting, on the ground, or even for choking self-rescue practice with a counter or chair. Includes 10 foam plugs and convenient carrying bag for on-the-go training sessions.