Luciferase Assay Kits, Biotium
Supplier: Biotium
30028-1 30028-2 30028-3 30028-T 99923 30028-L1 30028-L2 30028-L3
89138-958EA 221.29 CAD
89138-958 89138-960 89138-962 89138-964 89410-734 10018-364 10018-366 10018-368
Luciferase Assay Kits, Biotium
Assays Enzyme Assays
The Firefly Luciferase, Renilla Luciferase, and Dual Luciferase assays are flash-type assays for single sample luminometers or luminescence plate readers with sample injection systems
  • Flash-type assays for single sample luminometers or luminescence plate readers with sample injection systems
  • Steady-Luc assay three hour luminescence half-life
  • Lyophilized kits have assay buffers provided as lyophilized solids
The Firefly Luciferase, Renilla Luciferase, and Dual Luciferase assays are flash-type assays for single sample luminometers or luminescence plate readers with sample injection systems. The Steady-Luc Assay is a glow-type assay with a luminescence half-life of about three hours. The Steady-Luc kit can be used with luminescence plate readers without sample injection systems or for high-throughput screening. Lyophilized kits have assay buffers provided as lyophilized solids, for room temperature shipping and convenient storage.
The Steady-Luc Assay is a glow-type assay with a luminescence half-life of about 3 hours. The Steady-Luc kit can be used with luminescence plate readers without sample injection systems or for high-throughput screening. Lyophilized kits have assay buffers provided as lyophilized solids, for room temperature shipping and convenient storage.
Ordering information: Biotium also offers catalog number ATP-Glo™ Bioluminometric Cell Viability Assay (catalog number 89138-938).