Pierce™ Rapid Equilbrium Dialysis (RED) Device
Supplier: Thermo Scientific
90006 90007 90112 91012 99006
CAPI90006PK 2263.48 CAD
CAPI90006 CAPI90007 CAPI90112 CAPI91012 CAPI99006
Pierce™ Rapid Equilbrium Dialysis (RED) Device
Tubes Dialysis Tubes
Single-Use RED (rapid equilibrium dialysis) Plate is composed of disposable high-density polypropylene and is pre-loaded with 48 equilibrium dialysis membrane inserts. Each insert is comprised of two side-by-side chambers separated by an O-ring-sealed vertical cylinder of dialysis membrane with varying molecular-weight cutoffs (8 or 12 kD MWCO).
  • Easy and ready to use
  • Design enables equilibrium to reach 200 min⁻¹ agitation with in three to four hours
  • Reproducible and accurate for plasma binding assays
  • Robust compartmentalised design eliminates potential for
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable