Potassium iodide for biochemistry

Supplier: G-Biosciences

Synonyms: Dipotassium diiodide, Kali iodide

RC-151 RC-152
CA71003-538EA 649.29 CAD
CA71003-538 CA71003-540
Potassium iodide for biochemistry
Potassium iodide
Formula: Kl
Boiling Pt: 1330 °C (1013 hPa)
Melting Pt: 686 °C
Density: 3.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR