Western ReProbe™ Stripping Solution, G-Biosciences
Supplier: G-Biosciences
786-119 786-305 786-306 786-307 786-308 786-309
CA82022-512EA 647.71 CAD
CA82022-512 CA71003-118 CA71003-120 CA71003-122 CA71003-124 CA71003-126
Western ReProbe™ Stripping Solution, G-Biosciences
Electrophoresis Reagents Western Blotting Reagents
G-Biosciences' Western ReProbe™ is a single component system, specifically formulated to dissociate and remove antibodies from membrane bound proteins without destroying the antigenic binding affinity and does not use denaturants, SDS or boiling
G-Biosciences' Western ReProbe™ PLUS is a modified formulation of their Western ReProbe™ that allows for the removal of stubborn, high affinity antibodies from membrane bound proteins without destroying the antigenic binding affinity.
Western ReProbe™ and Western ReProbe™ PLUS are not recommended for stripping color producing Western blots that use substrates such as TMB, chloronapthol and DAB. Require no dilution; supplied as 5X solution; use 15-20 mL for each standard (7.5 x 8.5 cm) Western blot.
Western ReProbe™ allows you the ability to reuse your Western blots. The stripped blots can then be probed with new probes.
Ordering information: Accessories available.